COMH4211B85F Combi-Blocks, Inc.

(9H-Fluoren-9-yl)methyl 4-bromophenethylcarbamate

  • MDL number MFCD28053459
  • Cas Number : 1344158-44-6
  • Molecular Weight : 422.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5240

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5240
Availability details for
Quantity
  • Availability
  • Add to Cart