COMH4211B860 Combi-Blocks, Inc.

Methyl 3-((6-bromopyridin-3-yloxy)methyl)benzoate

  • MDL number MFCD11874520
  • Cas Number : 1121627-56-2
  • Molecular Weight : 322.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5242

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5242
Availability details for
Quantity
  • Availability
  • Add to Cart