COMH4211B862 Combi-Blocks, Inc.

(E)-3-(1H-Indol-6-yl)-2-propenoic acid methyl ester

  • MDL number MFCD20482894
  • Cas Number : 119160-35-9
  • Molecular Weight : 201.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5247

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5247
Availability details for
Quantity
  • Availability
  • Add to Cart