COMH4211B865 Combi-Blocks, Inc.

1-(2-Ethoxy-3-methyl-phenyl)-ethylamine hydrochloride

  • MDL number MFCD31557896
  • Cas Number : 2204052-21-9
  • Molecular Weight : 215.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5253

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5253
Availability details for
Quantity
  • Availability
  • Add to Cart