COMH4211B867 Combi-Blocks, Inc.

Trans-methyl 4-(methylamino)cyclohexanecarboxylate hydrochloride

  • MDL number MFCD28383636
  • Cas Number : 146447-21-4
  • Molecular Weight : 207.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5256

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5256
Availability details for
Quantity
  • Availability
  • Add to Cart