COMH4211B869 Combi-Blocks, Inc.

4-(4-Ethoxyphenyl)butanoic acid

  • MDL number MFCD00662535
  • Cas Number : 58472-30-3
  • Molecular Weight : 208.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5260

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5260
Availability details for
Quantity
  • Availability
  • Add to Cart