COMH4211B86A Combi-Blocks, Inc.

2,3,5-Trichloro-n-(2-oxo-ethyl)-benzamide

  • MDL number MFCD31381665
  • Cas Number : 477772-60-4
  • Molecular Weight : 266.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5261

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5261
Availability details for
Quantity
  • Availability
  • Add to Cart