COMH4211B86C Combi-Blocks, Inc.

[4-(4-Bromo-phenyl)-piperazin-1-yl]-acetic acid benzyl ester

  • MDL number MFCD31583319
  • Cas Number : 2270907-79-2
  • Molecular Weight : 389.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5265

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5265
Availability details for
Quantity
  • Availability
  • Add to Cart