COMH4211B86D Combi-Blocks, Inc.

2-Amino-5-fluoro-n-propylbenzamide

  • MDL number MFCD11132581
  • Cas Number : 1094449-17-8
  • Molecular Weight : 196.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-5-fluoro-N-propylbenzamide
Inchi Code 1S/C10H13FN2O/c1-2-5-13-10(14)8-6-7(11)3-4-9(8)12/h3-4,6H,2,5,12H2,1H3,(H,13,14)
Inchi Key UWACAROKGRLBGW-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5266

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5266
Availability details for
Quantity
  • Availability
  • Add to Cart