COMH4211B86E Combi-Blocks, Inc.

4-Bromo-3-methyl-n-(prop-2-en-1-yl)benzamide

  • MDL number MFCD26802223
  • Cas Number : 1566892-86-1
  • Molecular Weight : 254.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5268

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5268
Availability details for
Quantity
  • Availability
  • Add to Cart