COMH4211B86F Combi-Blocks, Inc.

2-[(5-Bromo-2-nitrophenyl)amino]ethan-1-ol

  • MDL number MFCD21193896
  • Cas Number : 854227-37-5
  • Molecular Weight : 261.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5271

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5271
Availability details for
Quantity
  • Availability
  • Add to Cart