COMH4211B870 Combi-Blocks, Inc.

3-Amino-n-benzyl-2-methylbenzamide

  • MDL number MFCD09812946
  • Cas Number : 301527-51-5
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5272

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5272
Availability details for
Quantity
  • Availability
  • Add to Cart