COMH4211B871 Combi-Blocks, Inc.

N,N-Dicyclohexyl-3-methyl-2-butenamide

  • MDL number MFCD28383618
  • Cas Number : 15745-13-8
  • Molecular Weight : 263.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5273

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5273
Availability details for
Quantity
  • Availability
  • Add to Cart