COMH4211B873 Combi-Blocks, Inc.

Cyclopropyl-(4-methoxy-2-trifluoromethyl-benzyl)-amine

  • MDL number MFCD31614072
  • Molecular Weight : 245.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5278

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5278
Availability details for
Quantity
  • Availability
  • Add to Cart