COMH4211B877 Combi-Blocks, Inc.

3-(4-Chlorophenyl)-1,2-oxazole-4-carboxylic acid

  • MDL number MFCD11053212
  • Cas Number : 1048922-51-5
  • Molecular Weight : 223.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5287

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5287
Availability details for
Quantity
  • Availability
  • Add to Cart