COMH4211B87C Combi-Blocks, Inc.

5-Bromo-2-cyclopropylmethoxy-benzoic acid

  • MDL number MFCD12138844
  • Cas Number : 1154385-13-3
  • Molecular Weight : 271.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5299

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5299
Availability details for
Quantity
  • Availability
  • Add to Cart