COMH4211B87D Combi-Blocks, Inc.

4-(3-Methylbutyl)benzoic acid

  • MDL number MFCD20644179
  • Cas Number : 105337-37-9
  • Molecular Weight : 192.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5300

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5300
Availability details for
Quantity
  • Availability
  • Add to Cart