COMH4211B87E Combi-Blocks, Inc.

5-Bromo-3,4-dihydro-1h-isoquinoline-2-carboxylic acid methylamide

  • MDL number MFCD31613768
  • Cas Number : 2123572-96-1
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5304

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5304
Availability details for
Quantity
  • Availability
  • Add to Cart