COMH4211B881 Combi-Blocks, Inc.

4-Bromo-2-methoxy-1-(2,2,2-trifluoro-ethoxy)-benzene

  • MDL number MFCD31539510
  • Cas Number : 1872891-66-1
  • Molecular Weight : 285.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5307

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5307
Availability details for
Quantity
  • Availability
  • Add to Cart