COMH4211B883 Combi-Blocks, Inc.

Methyl 5-(pyridin-4-yl)furan-2-carboxylate

  • MDL number MFCD31583066
  • Cas Number : 216867-36-6
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5313

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5313
Availability details for
Quantity
  • Availability
  • Add to Cart