COMH4211B884 Combi-Blocks, Inc.

(R)-5-Bromo-2-((tetrahydrofuran-3-yl)oxy)benzonitrile

  • MDL number MFCD31613966
  • Cas Number : 1893403-92-3
  • Molecular Weight : 268.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5315

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5315
Availability details for
Quantity
  • Availability
  • Add to Cart