COMH4211B886 Combi-Blocks, Inc.

Trans-4-morpholin-4-ylmethyl-cyclohexylamine hydrochloride

  • MDL number MFCD28556945
  • Cas Number : 2365390-54-9
  • Molecular Weight : 234.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5320

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5320
Availability details for
Quantity
  • Availability
  • Add to Cart