COMH4211B887 Combi-Blocks, Inc.

Methyl 7-iodoheptanoate

  • MDL number MFCD25962958
  • Cas Number : 38315-25-2
  • Molecular Weight : 270.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5323

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5323
Availability details for
Quantity
  • Availability
  • Add to Cart