COMH4211B88A Combi-Blocks, Inc.

Carbamic acid, n-methoxy-n-2-ethanol-1-yl-, 1,1-dimethylethyl ester

  • MDL number MFCD30729371
  • Cas Number : 1782512-07-5
  • Molecular Weight : 191.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5327

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5327
Availability details for
Quantity
  • Availability
  • Add to Cart