COMH4211B88B Combi-Blocks, Inc.

1-[2-(2-Fluorophenyl)ethyl]piperazine dihydrochloride

  • MDL number MFCD21090446
  • Cas Number : 1266694-49-8
  • Molecular Weight : 281.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5330

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5330
Availability details for
Quantity
  • Availability
  • Add to Cart