COMH4211B891 Combi-Blocks, Inc.

4-[4-(tert-Butoxycarbonyl)piperazin-1-yl]-2-fluorobenzoic acid

  • MDL number MFCD11872516
  • Cas Number : 1121596-45-9
  • Molecular Weight : 324.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5345

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5345
Availability details for
Quantity
  • Availability
  • Add to Cart