COMH4211B893 Combi-Blocks, Inc.

4-Amino-2-fluoro-n,n-dimethylbenzamide

  • MDL number MFCD16084618
  • Cas Number : 1187368-67-7
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5361

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5361
Availability details for
Quantity
  • Availability
  • Add to Cart