COMH4211B895 Combi-Blocks, Inc.

3-(4-Ethoxy-phenyl)-butyric acid

  • MDL number MFCD11172897
  • Cas Number : 6555-32-4
  • Molecular Weight : 208.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5367

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5367
Availability details for
Quantity
  • Availability
  • Add to Cart