COMH4211B897 Combi-Blocks, Inc.

3-(1-Hydroxy-cyclobutyl)-phenol

  • MDL number MFCD30717649
  • Cas Number : 1888730-77-5
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5372

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5372
Availability details for
Quantity
  • Availability
  • Add to Cart