COMH4211B898 Combi-Blocks, Inc.

tert-Butyl 2-(4-aminophenoxy)acetate

  • MDL number MFCD11045794
  • Cas Number : 167843-57-4
  • Molecular Weight : 223.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5373

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5373
Availability details for
Quantity
  • Availability
  • Add to Cart