COMH4211B89B Combi-Blocks, Inc.

tert-Butyl 4-[(4-bromo-2-methylphenyl)methyl]piperazine-1-carboxylate

  • MDL number MFCD28967707
  • Cas Number : 1446819-44-8
  • Molecular Weight : 369.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5376

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5376
Availability details for
Quantity
  • Availability
  • Add to Cart