COMH4211B89C Combi-Blocks, Inc.

N,N,2-Trimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

  • MDL number MFCD18732734
  • Cas Number : 1011731-99-9
  • Molecular Weight : 289.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5377

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5377
Availability details for
Quantity
  • Availability
  • Add to Cart