COMH4211B89D Combi-Blocks, Inc.

3-Methoxy-4'-(trifluoromethyl)biphenyl-4-carbonitrile

  • MDL number MFCD18425308
  • Cas Number : 1261742-02-2
  • Molecular Weight : 277.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-methoxy-4'-(trifluoromethyl)[1,1'-biphenyl]-4-carbonitrile
Inchi Code 1S/C15H10F3NO/c1-20-14-8-11(2-3-12(14)9-19)10-4-6-13(7-5-10)15(16,17)18/h2-8H,1H3
Inchi Key XUCIGNUYBDGATA-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5378

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5378
Availability details for
Quantity
  • Availability
  • Add to Cart