COMH4211B89F Combi-Blocks, Inc.

(4-((1-Ethoxy-2-methyl-1-oxopropan-2-yl)oxy)phenyl)boronic acid pinacol ester

  • MDL number MFCD22494206
  • Cas Number : 1146546-99-7
  • Molecular Weight : 334.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5382

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5382
Availability details for
Quantity
  • Availability
  • Add to Cart