COMH4211B8A0 Combi-Blocks, Inc.

4-Acetyl-3-fluoro-benzoic acid benzyl ester

  • MDL number MFCD31613848
  • Cas Number : 2270905-99-0
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5385

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5385
Availability details for
Quantity
  • Availability
  • Add to Cart