COMH4211B8A4 Combi-Blocks, Inc.

4,4-Difluoro-1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenylsulfonyl)piperidine

  • MDL number MFCD31583301
  • Cas Number : 1628016-72-7
  • Molecular Weight : 387.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5395

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5395
Availability details for
Quantity
  • Availability
  • Add to Cart