COMH4211B8A5 Combi-Blocks, Inc.

2-(Cyclopropylmethoxy)-5-methylaniline

  • MDL number MFCD11645606
  • Cas Number : 1094717-64-2
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5400

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5400
Availability details for
Quantity
  • Availability
  • Add to Cart