COMH4211B8A7 Combi-Blocks, Inc.

2-(2-Methylamino-ethoxy)-isoindole-1,3-dione hydrochloride

  • MDL number MFCD31579348
  • Cas Number : 1261131-66-1
  • Molecular Weight : 256.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5404

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5404
Availability details for
Quantity
  • Availability
  • Add to Cart