COMH4211B8A8 Combi-Blocks, Inc.

1-(2-Isobutoxy-5-methyl-phenyl)-ethylamine

  • MDL number MFCD11053244
  • Cas Number : 1048917-08-3
  • Molecular Weight : 207.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5406

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5406
Availability details for
Quantity
  • Availability
  • Add to Cart