COMH4211B8AB Combi-Blocks, Inc.

1-(2-Bromoethoxy)-2-ethylbenzene

  • MDL number MFCD02030511
  • Cas Number : 915922-20-2
  • Molecular Weight : 229.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5414

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5414
Availability details for
Quantity
  • Availability
  • Add to Cart