COMH4211B8AC Combi-Blocks, Inc.

4-Isoxazolemethanol, 3-(4-chloro-2-fluorophenyl)-5-methyl-

  • MDL number MFCD22370403
  • Cas Number : 1159602-72-8
  • Molecular Weight : 241.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5417

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5417
Availability details for
Quantity
  • Availability
  • Add to Cart