COMH4211B8AD Combi-Blocks, Inc.

3-Chloro-4-(propane-2-sulfonyl)aniline

  • MDL number MFCD24063365
  • Cas Number : 1524969-32-1
  • Molecular Weight : 233.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5419

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5419
Availability details for
Quantity
  • Availability
  • Add to Cart