COMH4211B8AE Combi-Blocks, Inc.

2-Chloro-n-(3-iodo-4-methylphenyl)pyridine-4-carboxamide

  • MDL number MFCD13239307
  • Cas Number : 585544-17-8
  • Molecular Weight : 372.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5420

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5420
Availability details for
Quantity
  • Availability
  • Add to Cart