COMH4211B8AF Combi-Blocks, Inc.

2-Benzyloxy-2'-trifluoromethyl-biphenyl-4-carboxylic acid

  • MDL number MFCD31613927
  • Cas Number : 1355680-15-7
  • Molecular Weight : 372.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5423

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5423
Availability details for
Quantity
  • Availability
  • Add to Cart