COMH4211B8B0 Combi-Blocks, Inc.

3-Bromo-n-cyclopropyl-2-methylaniline

  • MDL number MFCD30185856
  • Cas Number : 245765-58-6
  • Molecular Weight : 226.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5424

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5424
Availability details for
Quantity
  • Availability
  • Add to Cart