COMH4211B8B1 Combi-Blocks, Inc.

1-Bromo-3-(difluoromethoxy)-5-(trifluoromethyl)benzene

  • MDL number MFCD22580767
  • Cas Number : 1417567-27-1
  • Molecular Weight : 291.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5425

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5425
Availability details for
Quantity
  • Availability
  • Add to Cart