COMH4211B8B2 Combi-Blocks, Inc.

4-Acetyl-piperazine-1,2-dicarboxylic acid 1-tert-butyl ester

  • MDL number MFCD21606040
  • Cas Number : 939997-63-4
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5426

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5426
Availability details for
Quantity
  • Availability
  • Add to Cart