COMH4211B8B3 Combi-Blocks, Inc.

5-Bromo-4-methyl-2-(oxetan-3-yloxy)-pyrimidine

  • MDL number MFCD31583177
  • Cas Number : 1936371-29-7
  • Molecular Weight : 245.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5427

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5427
Availability details for
Quantity
  • Availability
  • Add to Cart