COMH4211B8B4 Combi-Blocks, Inc.

1-Bromo-2-(2-methoxyethyl)benzene

  • MDL number MFCD00092252
  • Cas Number : 39767-89-0
  • Molecular Weight : 215.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5429

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5429
Availability details for
Quantity
  • Availability
  • Add to Cart