COMH4211B8B5 Combi-Blocks, Inc.

5-Amino-n-ethyl-4-fluoro-2-methyl-benzamide

  • MDL number MFCD31579289
  • Cas Number : 1863868-73-8
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5431

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5431
Availability details for
Quantity
  • Availability
  • Add to Cart